missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, 18% in H{2}O, colloidal dispersion, stabilized with 1.3% yttrium oxide, Thermo Scientific Chemicals

Catalog No. p-4782251
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4012336 500 g
AA40123A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4012336 Supplier Thermo Scientific Chemicals Supplier No. 04012336
Only null left

Stabilized with 1.3% yttrium

Zirconium(IV) oxide is mainly employed in ceramic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O

Specifications

Physical Form Particles in Liquid
Quantity 500 g
Solubility Information Insoluble in water.
Chemical Name or Material Zirconium(IV) Oxide, Colloidal Dispersion
Hazard Category H290-H315-H318
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
EINECSNumber 215-227-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only