missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium(IV) Butoxide (ca. 80% in 1-Butanol), TCI America™
Description
- Concentration: 80.0 to 86.0 w/w%
- Physical State: Liquid
- Molecular Weight: 383.68 g/mol
- Molecular Formula: C16H36O4Zr
- Flash Point: 38°C
- Chemical Composition: Zirconium(IV) butoxide (ca. 80 to 86 w/w% in 1-butanol)
- Appearance: White to yellow to green clear liquid
- Specific Gravity: 1.06 (20/20°C)
- Storage Conditions: Store at room temperature (recommended in a cool and dark place, <15°C) under inert gas; moisture sensitive
Specifications
Specifications
| Color | Yellow |
| Physical Form | Liquid |
| Quantity | 500 g |
| Linear Formula | Zr(OC4H9)4 |
| UN Number | 1120 |
| Beilstein | 11535206 |
| Flash Point | 38°C (100.4°F) |
| Formula Weight | 383.68 |
| Concentration or Composition (by Analyte or Components) | Zirconium(IV) Butoxide: 80.0 to 86.0% w/w |
| Sensitivity | Moisture Sensitive |
| Show More |
Safety and Handling
| EINECSNumber | (2)-0226&(2)-2200 |
| TSCA | Yes |