missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) Butoxide (ca. 80% in 1-Butanol), TCI America™

Catalog No. Z001625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z001625G 25 g
Z0016500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z001625G Supplier TCI America Supplier No. Z001625G
Only null left

Chemical Identifiers

CAS 1071-76-7
Molecular Formula C16H40O4Zr+4
Molecular Weight (g/mol) 387.716
MDL Number MFCD00048765
InChI Key ASUYWGMDIAPAKI-UHFFFAOYSA-N
Synonym 1-butanol, zirconium 4+ salt 4:1, 1-butanol, zirconium 4++ salt
PubChem CID 122130442
IUPAC Name butan-1-ol;zirconium(4+)
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Zr+4]

Specifications

Color Yellow
Physical Form Liquid
Quantity 25 g
UN Number 1120
Formula Weight 383.68
Chemical Name or Material Zirconium(IV) Butoxide (ca. 80% in 1-Butanol)
EINECSNumber (2)-0226&(2)-2200
TSCA Yes