missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) bromide, 99%, Thermo Scientific Chemicals

Catalog No. p-4782250
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8613909 10 g
AA8613918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8613909 Supplier Thermo Scientific Chemicals Supplier No. 08613909

Zirconium(IV) bromide is used as a starting material in the synthesis of zirconium hydride complexes as well as unique cluster compounds. It is also used as a biochemical for proteomics research and as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13777-25-8
Molecular Formula Br4Zr
Molecular Weight (g/mol) 410.84
MDL Number MFCD00049637
InChI Key LSWWNKUULMMMIL-UHFFFAOYSA-J
Synonym zirconium tetrabromide, zrbr4, zirconium iv bromide, zirconium bromide zrbr4 , t-4, zirconium bromide zrbr4, zirconium bromide zrbr4 8ci 9ci, zirconium iv bromide, anhydrous trace metals basis
PubChem CID 83727
IUPAC Name tetrabromozirconium
SMILES [Br-].[Br-].[Br-].[Br-].[Zr+4]

Specifications

Melting Point 450° (15 atm)
Physical Form Powder
Quantity 10 g
Assay Percent Range 99%
Solubility Information Soluble in water.
Formula Weight 410.86
Sensitivity Moisture sensitive
Density 4.201
Chemical Name or Material Zirconium(IV) Bromide
Hazard Category H314-H335
Hazard Statement When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: III
EINECSNumber 237-417-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only