missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) 2-ethylhexanoate, 97%, Thermo Scientific Chemicals

$140.34 - $398.96

Chemical Identifiers

CAS 2233-42-3
Molecular Formula C32H60O8Zr
Molecular Weight (g/mol) 664.048
MDL Number MFCD00072684
InChI Key OFYFURKXMHQOGG-UHFFFAOYSA-J
Synonym zirconium tetra 2-ethylhexanoate, zirconium 2-ethylhexanoate, 2-ethylhexanoate; zirconium 4+, zirconium iv 2-ethylhexanoate, acmc-1cfv3, zirconium octoate, 2-ethylhexanoate; zirconium +4 cation, zirconium iv 2-ethylhexanoate 25g, tetrakis 2-ethylhexanoic acid zirconium iv salt, zirconium 4+ tetrakis 2-ethylhexanoate
PubChem CID 6451377
IUPAC Name 2-ethylhexanoate;zirconium(4+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zr+4]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3917414
View Documents
Thermo Scientific Chemicals
03917414
25 g
Each for $140.34
Only null left
 
AA3917422
View Documents
Thermo Scientific Chemicals
03917422
100 g
Each for $398.96
Only null left
 
Description

Description

Zirconium(IV) 2-ethylhexanoate is utilized as a metal organic precursor of zirconium for solution deposition and chemical vapor deposition of thin films containing zirconium especially zirconium oxide thin films. It is also used as yttria stabilized zirconia films for solid oxide fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2233-42-3
664.048
OFYFURKXMHQOGG-UHFFFAOYSA-J
6451377
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zr+4]
C32H60O8Zr
MFCD00072684
zirconium tetra 2-ethylhexanoate, zirconium 2-ethylhexanoate, 2-ethylhexanoate; zirconium 4+, zirconium iv 2-ethylhexanoate, acmc-1cfv3, zirconium octoate, 2-ethylhexanoate; zirconium +4 cation, zirconium iv 2-ethylhexanoate 25g, tetrakis 2-ethylhexanoic acid zirconium iv salt, zirconium 4+ tetrakis 2-ethylhexanoate
2-ethylhexanoate;zirconium(4+)

Specifications

2233-42-3
>93°C (199°F)
MFCD00072684
Moisture sensitive
OFYFURKXMHQOGG-UHFFFAOYSA-J
2-ethylhexanoate;zirconium(4+)
6451377
97%
Zirconium(IV) 2-ethylhexanoate
Amber
C32H60O8Zr
25 g
zirconium tetra 2-ethylhexanoate, zirconium 2-ethylhexanoate, 2-ethylhexanoate; zirconium 4+, zirconium iv 2-ethylhexanoate, acmc-1cfv3, zirconium octoate, 2-ethylhexanoate; zirconium +4 cation, zirconium iv 2-ethylhexanoate 25g, tetrakis 2-ethylhexanoic acid zirconium iv salt, zirconium 4+ tetrakis 2-ethylhexanoate
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zr+4]
664.048
664.04
Viscous Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P210-P281-P308+P313-P370+P378q-P501c

H227-H361

EINECSNumber : 218-776-6

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only