missing translation for 'onlineSavingsMsg'
Learn More

Zirconium ICP Standard, 10,000 ppm Zr in 5% HNO3, Ricca Chemical

Catalog No. PZR10KN100
Change view
Click to view available options
Package: Natural LDPE; Size: 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PZR10KN100 Package: Natural LDPE; Size: 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PZR10KN100 Supplier Ricca Chemical Company Supplier No. PZR10KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural LDPE Bottle
CAS 13826-66-9, 7697-37-2, 7732-18-5
Concentration or Composition Notes 10,000ppm Zr
CAS Min % 89.0, 6.68, 2.43
CAS Max % 93.0, 6.95, 2.53
Equivalence 1 mL = 10 mg Zr
Molecular Formula H2N2O7Zr
InChI Key UJVRJBAUJYZFIX-UHFFFAOYSA-N
SMILES [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
IUPAC Name nitric acid;oxozirconium
Molecular Weight (g/mol) 233.247
PubChem CID 83761
Chemical Name or Material Zirconium ICP Standard
Show More Show Less