missing translation for 'onlineSavingsMsg'
Learn More

Zirconium foil, 0.127mm (0.005in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4825008
Change view
Click to view available options
Quantity:
100 x 125 mm
125 x 500 mm
125 x 2000 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10594GM 100 x 125 mm
AA10594GR 125 x 500 mm
AA10594IB 125 x 2000 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10594GM Supplier Thermo Scientific Chemicals Supplier No. 010594GM
Only null left

Form: Approx.10.3g/100 x 125mm, Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Color Gray to Silver
Appearance Gray to silver foil
Quantity 100 x 125 mm
Odor Odorless
Form Foil
Assay Percent Range 99.5% (Metals basis)
Chemical Name or Material Zirconium, Annealed
EINECSNumber 231-176-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only