missing translation for 'onlineSavingsMsg'
Learn More

Zirconium foil, 0.025mm (0.001in) thick, annealed, 99.8% (metals basis excluding Hf)

Catalog No. AA38369VS
Change view
Click to view available options
Quantity:
10 x 50 cm
10 x 200 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38369VS 10 x 200 cm
AA38369VR 10 x 50 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38369VS Supplier Thermo Scientific Chemicals Supplier No. 038369VS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-67-7 | Zr | 91.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Quantity 10 x 200 cm
Odor Odorless
Weight ≈8.3g/10x50cm
Form Foil
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Zirconium foil, 0.025mm (0.001 in.) thick, Annealed, Temper: soft
EINECSNumber 231-176-9
RTECSNumber ZH7070000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only