missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium foil, 0.025mm (0.001in) thick, 99.9+% (metals basis)
CAS: 7440-67-7 | Zr | 91.22 g/mol
Supplier: Thermo Scientific Chemicals 043049FQ
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Zirconium foil | |
| 7440-67-7 | |
| Odorless | |
| Foil | |
| Zr | |
| zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
| [Zr] | |
| 91.22 | |
| CHEBI:33342 |
| 0.025mm (0.001 in.) thick | |
| 25 x 100 mm | |
| ≈0.1g/25x25mm | |
| (metals basis) | |
| MFCD00011303 MFCD00148878 | |
| QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
| zirconium | |
| 23995 | |
| ≥99.9% |
Chemical Identifiers
| 7440-67-7 | |
| 91.22 | |
| QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
| 23995 | |
| zirconium |
| Zr | |
| MFCD00011303 MFCD00148878 | |
| zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
| CHEBI:33342 | |
| [Zr] |
Safety and Handling
EINECSNumber : 231-176-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only