missing translation for 'onlineSavingsMsg'
Learn More

Zirconium foil, 0.025mm (0.001in) thick, 99.9+% (metals basis)

Catalog No. AA43049FW
Change view
Click to view available options
Quantity:
25 x 25 mm
25 x 100 mm
25 x 500 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43049FW 25 x 500 mm
AA43049FF 25 x 25 mm
AA43049FQ 25 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43049FW Supplier Thermo Scientific Chemicals Supplier No. 043049FW

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-67-7 | Zr | 91.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Quantity 25 x 500 mm
Odor Odorless
Weight ≈0.1g/25x25mm
Form Foil
Percent Purity ≥99.9%
Assay (metals basis)
Chemical Name or Material Zirconium foil, 0.025mm (0.001 in.) thick
EINECSNumber 231-176-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only