missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ ZipTip™ Pipet Tips
Sample preparation and precise liquid handling in one device
Supplier: MilliporeSigma™ ZTC18M008
Description
Sample preparation and precise liquid handling in one device
A 0.6μL bed of C18, C4, or SCX (strong cation exchange) media is fixed at the end of a 10μL pipet tip such that there is no dead volume; concentrated, purified samples are eluted in 1 to 4μL. For applications requiring smaller elution volumes (e.g., <1μL), ZipTip μ-C18 tips feature a microbed format containing 0.2μL of media.
- Concentrate and purify 1–100μL biological samples
- Ideal for purifying and concentrating femtomoles to picomoles of peptides, proteins, or oligonucleotides in minutes prior to mass spectrometry, HPLC, capillary electrophoresis, and other analytical techniques
Specifications
| 10 μL | |
| 3cm | |
| Specialty, Media-Fixed | |
| Universal Fit | |
| Polypropylene |
| 1.2 in. | |
| Micro Bed of C18 Media | |
| 8/Pk. | |
| ZipTip |