missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ZipTip™ Pipet Tips

Sample preparation and precise liquid handling in one device

Supplier:  MilliporeSigma™ ZTSCXS008

 View more versions of this product

Catalog No. ZTSCXS008


Only null left

Description

Description

Sample preparation and precise liquid handling in one device

A 0.6μL bed of C18, C4, or SCX (strong cation exchange) media is fixed at the end of a 10μL pipet tip such that there is no dead volume; concentrated, purified samples are eluted in 1 to 4μL. For applications requiring smaller elution volumes (e.g., <1μL), ZipTip μ-C18 tips feature a microbed format containing 0.2μL of media.

  • Concentrate and purify 1–100μL biological samples
  • Ideal for purifying and concentrating femtomoles to picomoles of peptides, proteins, or oligonucleotides in minutes prior to mass spectrometry, HPLC, capillary electrophoresis, and other analytical techniques

Order Info

Available in two packaging configurations: resealable trays (packs of 8 and 24 tips) and 96-place tip racks (packs of 96 and packs of 960 tips)

Specifications

Chemical Identifiers

10 μL
Universal Fit
ZipTip
Polypropylene

Specifications

10 μL
3cm
Specialty, Media-Fixed
Universal Fit
Polypropylene
1.2 in.
Strong Cation Exchange Media
8/Pk.
ZipTip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions