missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ZipTip™ Pipet Tips

Catalog No. ZTC18S096
Change view
Click to view available options
For Use With (Application):
C18 Media
C4 Media
Micro Bed of C18 Media
Strong Cation Exchange Media
Quantity:
8/Pk.
96/Pk.
960/Pk.
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Quantity
ZTC18S096 C18 Media 96/Pk.
ZTC04S008 C4 Media 8/Pk.
ZTC04S096 C4 Media 96/Pk.
ZTSCXS096 Strong Cation Exchange Media 96/Pk.
ZTSCXS008 Strong Cation Exchange Media 8/Pk.
ZTC18S960 C18 Media 960/Pk.
ZTC04S960 C4 Media 960/Pk.
ZTC18M960 Micro Bed of C18 Media 960/Pk.
ZTC18M096 Micro Bed of C18 Media 96/Pk.
ZTC18M008 Micro Bed of C18 Media 8/Pk.
ZTC18S008 C18 Media 8/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. ZTC18S096 Supplier MilliporeSigma™ Supplier No. ZTC18S096

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sample preparation and precise liquid handling in one device

A 0.6μL bed of C18, C4, or SCX (strong cation exchange) media is fixed at the end of a 10μL pipet tip such that there is no dead volume; concentrated, purified samples are eluted in 1 to 4μL. For applications requiring smaller elution volumes (e.g., <1μL), ZipTip μ-C18 tips feature a microbed format containing 0.2μL of media.

  • Concentrate and purify 1–100μL biological samples
  • Ideal for purifying and concentrating femtomoles to picomoles of peptides, proteins, or oligonucleotides in minutes prior to mass spectrometry, HPLC, capillary electrophoresis, and other analytical techniques

Order Info

Available in two packaging configurations: resealable trays (packs of 8 and 24 tips) and 96-place tip racks (packs of 96 and packs of 960 tips)

Specifications

Volume (Metric) 10 μL
Volume (Metric) Dispensing 10 μL
Filtered Filtered
Sterility Non-sterile
Length (English) 1.25 in.
Length (Metric) 3.1cm
Storage Requirements 4°C to 70°C
For Use With (Application) C18 Media
Tip Style Specialty, Media-Fixed
Quantity 96/Pk.
Product Type Pipette Tips
Content And Storage Store at 4°C to 70°C
Shipping Condition Ambient
Disposable Single-use
For Use With (Equipment) Universal Fit
Product Line ZipTip
Material Polypropylene
Show More Show Less