missing translation for 'onlineSavingsMsg'
Learn More

Ziprasidone 98.0+%, TCI America™

Catalog No. Z00341G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z00341G 1 g
Z0034100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z00341G Supplier TCI America Supplier No. Z00341G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 146939-27-7
Molecular Formula C21H21ClN4OS
Molecular Weight (g/mol) 412.936
MDL Number MFCD00866661
InChI Key MVWVFYHBGMAFLY-UHFFFAOYSA-N
PubChem CID 60854
ChEBI CHEBI:10119
IUPAC Name 5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one
SMILES C1CN(CCN1CCC2=C(C=C3C(=C2)CC(=O)N3)Cl)C4=NSC5=CC=CC=C54

Specifications

Melting Point 222°C
Quantity 1 g
Formula Weight 412.94
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ziprasidone
RTECSNumber NM3241000
TSCA No