Learn More
ZIP-MIX™ Plaster Casting Kit
Catalog No. S69042B
Change view
Click to view available options
ZIP-MIX Plaster Casting
Catalog No.
S69042B ZIP-MIX Plaster Casting
1 options
Catalog No. S69042B Supplier Customer Channels Group (Ccg) Supplier No. ZMP1005
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
24 oz. bottle of pretreated water, 3 lb. package of plaster and one 13 x 18 in. mixing bag
Eliminates the mess and difficulty of mixing plaster.
  • Ensures a uniform, smooth mixture every time — free of lumps
  • Simply combine water and plaster in the zip-top bag and knead with both hands until no lumps are felt
  • Everything needed to cast a full-sized adult footprint is in one convenient package
Specifications
For Use With (Application) For eliminate the mess and difficulty of mixing plaster
Includes One 24 oz. zmp101 bottle of pre-treated water, one 3 lb. zmp102 package of plaster, 1 - zte031 zip-mix™ mixing bag, 13 x 18 in.
Product Type Zip-Mix™ Plaster Casting Kit