Learn More
Abcam Inc Zinquin, Zn2+ selective fluorescent probe, 1MG
Catalog No. 502705182
Encompass
MW 386.4 Da, Purity >95%. Intracellular Zn2+ selective fluorescent probe. Forms zinquin-Zn2+ complexes in either 1:1 or 2:1 ratios (Kd values are 370 and 850 nM respectively). Zinquin ethyl ester (ab145478) analog. Labels micromolar concentrations of intracellular Zn2+ ions with an intense extranuclear fluorescence emitting in the blue range.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502705182 Supplier Abcam Inc Supplier No. AB1454511MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 386.4 Da, Purity >95%. Intracellular Zn2+ selective fluorescent probe. Forms zinquin-Zn2+ complexes in either 1:1 or 2:1 ratios (Kd values are 370 and 850 nM respectively). Zinquin ethyl ester (ab145478) analog. Labels micromolar concentrations of intracellular Zn2+ ions with an intense extranuclear fluorescence emitting in the blue range.
The product is subject to the following: Abcam Restricted Use Statement