Learn More
Abcam Inc Zinquin ethyl ester, Zn2+ selective fluorescent probe, 1MG
Catalog No. 502705168
Encompass
MW 414.5 Da, Purity >99%. Cell-permeable Zn2+ selective fluorescent probe. Zinquin (ab145451) precursor. Labels micromolar concentrations of intracellular Zn2+ ions with an intense extranuclear fluorescence emitting in the blue range.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502705168 Supplier Abcam Inc Supplier No. AB1454781MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 414.5 Da, Purity >99%. Cell-permeable Zn2+ selective fluorescent probe. Zinquin (ab145451) precursor. Labels micromolar concentrations of intracellular Zn2+ ions with an intense extranuclear fluorescence emitting in the blue range.
The product is subject to the following: Abcam Restricted Use Statement