missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zincon monosodium salt, For Spectrophotometric Det. of Cu and Zn, MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | 250°C to 260°C |
| CAS | 62625-22-3 |
| Linear Formula | C20H15N4 NaO6 S |
| Quantity | 1 g |
| Formula Weight | 462.41 |