missing translation for 'onlineSavingsMsg'
Learn More

Zincon monosodium salt, For Spectrophotometric Det. of Cu and Zn, MilliporeSigma™ Supelco™

Catalog No. 111014933
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014933 1 g
111014934 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014933 Supplier Merck Emd Millipore Supplier No. 964401G

Molar absorption coefficients and stability constants of Zincon metal complexes for determination of metal ions and bioinorganic applications. A. Kocyla, A. Pomorski, A. Krezel, Journal of Inorganic Biochemistry 176, 53-65, (2017).

Chemical Identifiers

MDL Number MFCD00064385
Synonym 2-Carboxy-2 ′-hydroxy-5 ′-sulfoformazyl-benzene monosodium salt; 2-[5-(2-Hydroxy-5-sulfophenyl)-3-phenyl-1-formazyl]benzoic acid monosodium salt

Specifications

Melting Point 250°C to 260°C
CAS 62625-22-3
Linear Formula C20H15N4 NaO6 S
Quantity 1 g
Formula Weight 462.41