missing translation for 'onlineSavingsMsg'
Learn More

Zinc wire, 2.0mm (0.08in) dia, 99.95% (metals basis)

Form: approx. 22.4g/m | CAS: 7440-66-6 | Zn | 65.38 g/mol

Supplier:  Thermo Scientific Chemicals 042706G5

 View more versions of this product

Catalog No. AA42706G5


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc
7.14g/cm3
Odorless
Wire
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
7440-66-6
5 m
99.95%, metal basis
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
CHEBI:30185

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-175-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only