missing translation for 'onlineSavingsMsg'
Learn More

Zinc wire, 0.25mm (0.01in) dia, 99.95% (metals basis)

Catalog No. AA42703G5
Change view
Click to view available options
Quantity:
5 m
25 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42703G5 5 m
AA42703G9 25 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42703G5 Supplier Thermo Scientific Chemicals Supplier No. 042703G5
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Form: approx. 5.60g/m | CAS: 7440-66-6 | Zn | 65.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Appearance Gray wire
Quantity 5 m
Odor Odorless
Form Wire
Assay Percent Range 99.95% (Metals basis)
Chemical Name or Material Zinc
EINECSNumber 231-175-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only