missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfide, 99.99%, (trace metal basis), < 10 microns

Catalog No. AC223655000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223655000 500 g Glass bottle
AC223650250 25 g Glass bottle
AC223651000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC223655000 Supplier Thermo Scientific Chemicals Supplier No. 223655000

CAS: 1314-98-3 | SZn | 97.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 1314-98-3
Molecular Formula SZn
Molecular Weight (g/mol) 97.44
MDL Number MFCD00011301
InChI Key DRDVZXDWVBGGMH-UHFFFAOYSA-N
Synonym zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
PubChem CID 14821
SMILES [S--].[Zn++]

Specifications

Color White to Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.99%
Linear Formula ZnS
Merck Index 15, 10360
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
IUPAC Name zinc(2+) sulfanediide
Formula Weight 97.44
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Zinc sulfide, 99.99%, <10 microns
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Hazard Statement GHS H Statement
Contact with acids liberates toxic gas.
Precautionary Statement GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-251-3
RTECSNumber ZH5400000
TSCA TSCA

RUO – Research Use Only