missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfide, 99.99% (metals basis)

Catalog No. AA4009109
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4009109 10 g
AA4009118 50 g
AA4009130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4009109 Supplier Thermo Scientific Chemicals Supplier No. 04009109

Used in Luminescent material, infra red optical material, semiconductor, as a pigment, used as a window for visible optics and infrared optics. | CAS: 1314-98-3 | SZn | 97.44 g/mol

Used in Luminescent material, infra red optical material, semiconductor, as a pigment, used as a window for visible optics and infrared optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-98-3
Molecular Formula SZn
Molecular Weight (g/mol) 97.44
MDL Number MFCD00011301
InChI Key DRDVZXDWVBGGMH-UHFFFAOYSA-N
Synonym zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
PubChem CID 14821
SMILES [S--].[Zn++]

Specifications

Melting Point 1700°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99% metal basis
Solubility Information soluble in acids,insoluble in water.
IUPAC Name sulfanylidenezinc
Formula Weight 97.43
Odor Odorless
Density 4.1
Chemical Name or Material Zinc sulfide
Show More Show Less
EINECSNumber 215-251-3
RTECSNumber ZH5400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only