missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Solution, 0.1M, Solstice

Catalog No. 6001787
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001787 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001787 Supplier Solstice Supplier No. 353921L
Only null left

Volumetric, 0.1 M ZnSO4

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

Chemical Identifiers

CAS 7733-02-0
Molecular Formula O4SZn
Molecular Weight (g/mol) 161.44
MDL Number MFCD00011302
InChI Key NWONKYPBYAMBJT-UHFFFAOYSA-L
Synonym zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
PubChem CID 24424
ChEBI CHEBI:35176
IUPAC Name zinc;sulfate
SMILES [Zn++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 L
Linear Formula ZnSO4
UN Number UN3082
Formula Weight 161.45g/mol
Chemical Name or Material Zinc sulfate solution
Hazard Statement P273-P280-P305 + P351 + P338-P337 + P313