missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate concentrate, For 1L standard solution, 0.1 M ZnSO4, Honeywell Fluka™

Catalog No. p-7122057
Change view
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001786 1 Ea.
60046058 6 x 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001786 Supplier Honeywell Chemicals Supplier No. 320471EA
Only null left

for 1L standard solution, 0.1 M ZnSO4

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7733-02-0
Molecular Formula O4SZn
Molecular Weight (g/mol) 161.44
MDL Number MFCD00011302
InChI Key NWONKYPBYAMBJT-UHFFFAOYSA-L
Synonym zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
PubChem CID 24424
ChEBI CHEBI:35176
IUPAC Name zinc;sulfate
SMILES [Zn++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 Ea.
Linear Formula ZnSO4
UN Number 3082
Formula Weight 161.45g/mol
Packaging Ampoule
Chemical Name or Material Zinc Sulfate Concentrate
Hazard Statement P273-P280-P305 + P351 + P338-P501
Precautionary Statement H318-H410