Learn More
Zinc stearate, ZnO 12.5-14%
CAS: 557-05-1 | C36H70O4Zn | 632.332 g/mol
Supplier: Thermo Scientific Chemicals 033238A1
| Quantity | 1 kg |
|---|
Description
Zinc stearate is used as a release agent for the manufacturing of rubber, polyurethane and polyester processing. It is a lubricant and thickening agent in cosmetic industry. It plays a vital role as an activator for rubber vulcanization. It acts as a phase transfer catalyst in the saponification of fats due to its lipophilic character. It is an active component in fanning powder, which is utilized to decrease the friction between cards.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 557-05-1 | |
| 632.332 | |
| XOOUIPVCVHRTMJ-UHFFFAOYSA-L | |
| 11178 | |
| CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2] |
| C36H70O4Zn | |
| MFCD00013031 | |
| zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat | |
| zinc;octadecanoate |
Specifications
| Zinc stearate | |
| 130°C | |
| C36H70O4Zn | |
| MFCD00013031 | |
| zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat | |
| XOOUIPVCVHRTMJ-UHFFFAOYSA-L | |
| zinc;octadecanoate | |
| 11178 | |
| Powder |
| 557-05-1 | |
| White | |
| 1 kg | |
| 14,10158 | |
| Insoluble in water,alcohol and ether; Soluble in benzeneSoluble in acids,aromatic and chlorinated hydrocarbons; Insoluble in water,alcohol and ether. | |
| CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2] | |
| 632.332 | |
| 632.48 |
Safety and Handling
H500
EINECSNumber : 209-151-9
RTECSNumber : WI4390000
TSCA : Yes
Recommended Storage : Ambient temperatures