missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc stearate
CAS: 557-05-1 | C36H70O4Zn | 632.34 g/mol
Supplier: Thermo Scientific Chemicals 263780050
| Quantity | 5 kg |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 557-05-1 | |
| XOOUIPVCVHRTMJ-UHFFFAOYSA-L | |
| 11178 | |
| CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2] |
| MFCD00013031 | |
| zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat | |
| zinc;octadecanoate |
Specifications
| Zinc stearate | |
| 118.0°C to 128.0°C | |
| >100°C | |
| Fiber Drum | |
| [CH3(CH2)16CO2]2Zn | |
| MFCD00013031 | |
| zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat | |
| XOOUIPVCVHRTMJ-UHFFFAOYSA-L | |
| zinc;octadecanoate | |
| 11178 | |
| 12.5 to 14.0% (ZnO) |
| 557-05-1 | |
| White | |
| Authentic | |
| C36H70O4Zn | |
| 5 kg | |
| 15, 10358 | |
| Solubility in water: insoluble. Other solubilities: insoluble in ethanol,diethyl ether,,soluble in benzene,xylene,fats,oils | |
| CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2] | |
| 632.34 | |
| 632.34 | |
| Powder |
Safety and Handling
EINECSNumber : 209-151-9
RTECSNumber : ZH5200000
TSCA : TSCA