missing translation for 'onlineSavingsMsg'
Learn More

Zinc Stearate, Powder, USP, 12.5-14%, Spectrum™ Chemical

Catalog No. 18607876
Change view
Click to view available options
Quantity:
500 g
12 kg
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607876 500 g Poly Bottle
18607878 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18607876 Supplier Spectrum Chemical Mfg Cor Supplier No. ZI115500GM

ZI115, 557-05-1

Spectrum™ Chemical Zinc Stearate, Powder, USP is a mild antiseptic and astringent used as a local soothing application for inflammatory and irritating skin diseases and in pharmacuetical formulations as a lubricant in tablet and capsule manufacture. All Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 557-05-1
Molecular Formula C36H70O4Zn
Molecular Weight (g/mol) 632.33
InChI Key XOOUIPVCVHRTMJ-UHFFFAOYSA-L
IUPAC Name zinc(2+) dioctadecanoate
SMILES [Zn++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 12.5 to 14%
Grade USP
Packaging Poly Bottle