missing translation for 'onlineSavingsMsg'
Learn More

Zinc shot, 10mm (0.4in) dia x 2mm (0.08in) thick, 99.99% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol

$65.32 - $330.11

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1044022
View Documents
Thermo Scientific Chemicals
01044022
100 g
Each for $65.32
Only null left
 
AA1044036
View Documents
Thermo Scientific Chemicals
01044036
500 g
Each for $160.29
Only null left
 
AA10440A3
View Documents
Thermo Scientific Chemicals
010440A3
2 kg
Each for $330.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]

Specifications

Gray
Gray shot
Odorless
Shot
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
Zinc
7440-66-6
100 g
99.99% (Metals basis)
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
CHEBI:30185
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-175-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only