missing translation for 'onlineSavingsMsg'
Learn More

Zinc selenide

Catalog No. AAA1994709
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1994709 10 g
AAA1994718 50 g
AAA1994730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1994709 Supplier Thermo Scientific Chemicals Supplier No. A1994709

CAS: 1315-09-9 | SeZn | 144.35 g/mol

Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc selenide is used to form II-VI light-emitting diodes and diode lasers. It is doped with chromium (ZnSe:Cr) has been used as an infrared laser gain medium. It is used as an infrared optical material. It is used in x-ray and gamma ray detectors.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 1315-09-9
Molecular Formula SeZn
Molecular Weight (g/mol) 144.35
MDL Number MFCD00011296
InChI Key GTLQJUQHDTWYJC-UHFFFAOYSA-N
Synonym zinc selenide znse, znse, zinc selenide, spectro, zinc selenide, powder, 10 mum trace metals basis, zinc selenide crystal optic disc, 13mm x 1mm, polished both sides, zinc selenide crystal optic disc, 13mm x 2mm, polished both sides, zinc selenide crystal optic disc, 25mm x 2mm, polished both sides, zinc selenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick, zinc selenide optical window, 12.7mm 0.5in dia x 2mm 0.08in thick, zinc selenide optical window, 25.4mm 1.0in dia x 1mm 0.04in thick
IUPAC Name zinc(2+) selandiide
SMILES [Zn++].[Se--]

Specifications

Melting Point >1100°C
Quantity 10 g
UN Number UN3283
Merck Index 14,10156
Solubility Information Insoluble in water.
Formula Weight 144.33
Odor Odorless
Density 5.42^1^5
Chemical Name or Material Zinc selenide
Hazard Category H301-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P301+P310-P314-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 215-259-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only