missing translation for 'onlineSavingsMsg'
Learn More

Zinc selenide crystal optic disc, 13mm x 1mm, polished both sides

Catalog No. AA41883KT
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41883KT 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41883KT Supplier Thermo Scientific Chemicals Supplier No. 041883KT
Only null left

 

Spectroscopy, IR window

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Zinc selenide crystal optic disc
Name Note 13mm x 1mm, Polished both sides
Quantity 1 Ea.
MDL Number MFCD00011296

RUO – Research Use Only