missing translation for 'onlineSavingsMsg'
Learn More

Zinc rod, 10mm (0.39in) dia, 99.9998% (metals basis)

CAS: 7440-66-6 | Zn | 65.38 g/mol

$261.26 - $909.14

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA12718DJ
View Documents
Thermo Scientific Chemicals
012718DJ
50 mm
Each for $261.26
Only null left
 
AA12718DT
View Documents
Thermo Scientific Chemicals
012718DT
250 mm
Each for $909.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]

Specifications

7440-66-6
Odorless
Rod
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
99.9998%
(metals basis)
50 mm
≈5.60g/cm
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
CHEBI:30185
Zinc rod, 10mm (0.39 in.) dia.
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-175-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only