missing translation for 'onlineSavingsMsg'
Learn More

Zinc Powder, For Analysis, Particle Size < 45μm, EMSURE™, MilliporeSigma™

Catalog No. M1087890500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1087890500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1087890500 Supplier Technidata Supplier No. 1087890500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Appearance Silver granules
Quantity 500 g
Odor Odorless
Packaging Metal can
Vapor Pressure 1.33 hPa at 487°C
Form Solid
Formula Weight 65.39
Chemical Name or Material Zinc