missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc powder, average 5-8 micron, 97.5% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | Zinc |
| Couleur | Gray |
| Aspect | Poudre grise |
| Densité | 7.14 |
| Quantité | 100 g |
| Odeur | Inodore |
| Sensibilité | Sensible à l’air et à l’humidité |
| Fourchette de pourcentages d’essai | 97,5% (base des métaux) |
| Forme | Poudre/Flocons |
Sécurité et manipulation
| Hazard Category | H250-H260-H500 |
| Precautionary Statement | P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c |
| DOTInformation | DOT Class: 4.3, Packaging Group: II |
| EINECSNumber | 231-175-3 |
| RTECSNumber | ZG8600000 |
| TSCA | Oui |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only