missing translation for 'onlineSavingsMsg'
Learn More

Zinc peroxide, approximately 50% ZnO{2}

Catalog No. p-7050887
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3963522 100 g
AA3963536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3963522 Supplier Thermo Scientific Chemicals Supplier No. 03963522
Only null left

Curing agent, rubber accelerator | CAS: 1314-22-3 | O2Zn | 97.38 g/mol

Zinc peroxide is used as curing agent, rubber accelerator. has been applied in a variety of settings, from medicine to aesthetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-22-3
Molecular Formula O2Zn
Molecular Weight (g/mol) 97.38
MDL Number MFCD00137469
InChI Key IPTOGCUGCFHDSS-UHFFFAOYSA-N
Synonym zinc peroxide, zinc superoxide, zinc dioxide, zinc peroxide un1516 oxidizer, zinc peroxide, medicinal, unii-0i969dvm77, zinc peroxide, medicinal usp, zpo
PubChem CID 14809
SMILES [Zn++].[O-][O-]

Specifications

Melting Point >150°C (decomposition)
Physical Form Powder
Quantity 100 g
UN Number UN1516
Merck Index 14,10149
Solubility Information Insoluble in water.
IUPAC Name zinc(2+) dioxidanediide
Formula Weight 97.38
Concentration or Composition (by Analyte or Components) ≈50% ZnO2
Odor Odorless
Chemical Name or Material Zinc peroxide
Show More Show Less
Hazard Category H271
Hazard Statement GHS H Statement
H272-H332-H315-H319-H335
May intensify fire
oxidizer.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P280-P283-P306+P360-P370+P378q-P371+P380+P375-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: ZINC PEROXIDE
EINECSNumber 215-226-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only