missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc peroxide, approximately 50% ZnO{2}
Description
Zinc peroxide is used as curing agent, rubber accelerator. has been applied in a variety of settings, from medicine to aesthetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >150°C (decomposition) |
| Physical Form | Powder |
| Quantity | 500 g |
| UN Number | UN1516 |
| Merck Index | 14,10149 |
| Solubility Information | Insoluble in water. |
| IUPAC Name | hydrogen peroxide;zinc |
| Formula Weight | 97.38 |
| Concentration or Composition (by Analyte or Components) | ≈50% ZnO2 |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H271 |
| Hazard Statement | GHS H Statement H272-H332-H315-H319-H335 May intensify fire oxidizer. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P220-P221-P280-P283-P306+P360-P370+P378q-P371+P380+P375-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: ZINC PEROXIDE |
| EINECSNumber | 215-226-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only