missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.9% (metals basis), Thermo Scientific™

Catalog No. AA41126KS
Change view
Click to view available options
Quantity:
1 ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41126KS 1 ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41126KS Supplier Thermo Scientific Chemicals Supplier No. 41126KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.379
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Quantity 1 ea.
Solubility Information Soluble in diluted acid. Insoluble in water and alcohol.
Formula Weight 81.37
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Zinc oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 215-222-5
TSCA Yes
Recommended Storage Ambient temperatures