missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, Puratronic™, 99.9995% (metals basis)

Catalog No. p-4904576
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1285514 25 g
AA1285522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1285514 Supplier Thermo Scientific Chemicals Supplier No. 01285514
Only null left

CAS: 1314-13-2 | OZn | 81.38 g/mol

Zinc oxide is used as a sun screener. It is serves as an additive in products such as cigarette filters, plastics, anticorrosive coatings, ceramics, glass, cement, lubricants, ointments, adhesives, sealants, pigments, batteries, ferrites, fire retardants, and first-aid tapes. Zinc oxide plays an important role in textiles as UV-absorber. It can protect rubber from fungi and UV light. As a wide-bandgap semiconductor with favorable properties, it has the potential for use in transparent electrodes, in nanorod sensors, in liquid crystal displays, as thin-film transistors and light emitting diodes, and in heat-protecting windows.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9995% metal basis
Solubility Information Insoluble in water. Soluble in dilute acetic acid,and dilute mineral acids.
Formula Weight 81.37
Odor Odorless
Density 5.606
Chemical Name or Material Zinc oxide
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 215-222-5
RTECSNumber ZH4810000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only