missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide, Powder, ACS, 99%, Spectrum™ Chemical

$1084.22 - $1084.22

Chemical Identifiers

CAS 1314-13-2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614788
View Documents
Spectrum Chemical Mfg Cor
Z108025KG
2.5 kg Poly Bottle
Each for $1,084.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1314-13-2

Specifications

1314-13-2
2.5 kg
ACS
1
99%
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.