missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, NanoArc™ ZN-0625, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant

Catalog No. p-7052201
Change view
Click to view available options
Quantity:
100 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4492422 100 g
AA44924A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4492422 Supplier Thermo Scientific Chemicals Supplier No. 04492422
Only null left

Colloidal dispersion with dispersant | CAS: 1314-13-2 | OZn | 81.38 g/mol

Zinc oxide dispersion in Dowanol PMA suitable for incorporation in a wide variety of coatings formulations

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Zinc oxide dispersion in Dowanol™ PMA suitable for incorporation in a wide variety of coatings formulations

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Physical Form Powder
Quantity 100 g
Assay Percent Range 50% in 1, 2-propanediol monomethyl ether acetate
UN Number UN1993
Formula Weight 81.37
Odor Ester-like
Chemical Name or Material Zinc oxide, NanoTek™ Z1102PMA, 50% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion with dispersant
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only