missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, NanoArc™ ZN-0605

Catalog No. AA4584922
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4584922 100 g
AA4584936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4584922 Supplier Thermo Scientific Chemicals Supplier No. 04584922

CAS: 1314-13-2 | OZn | 81.38 g/mol

Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis. Used in electronic industry, instrumental industry, manufacture of electrical device, image recorder, rheostat, phosphor. Used as a sun screening agent in cosmetics, antibacterial and health protection antiager; UV protection; Piezoelectrics; High-temperature lubricant in gas turbine engines; Flame retardant; Adsorption; Dental cements; Environmental remediation; Gas sensors; Photocatalytic decontamination; Attenuation of ultraviolet light; Demilitarization of chemical and biological warfare agents; Cosmetics and cosmeceuticals; Electrodes for solar cells; Varistors; Pigments for paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula ZnO
UN Number UN3077
Merck Index 14,10147
Solubility Information Insoluble in water.
Formula Weight 81.37
Odor Odorless
Chemical Name or Material Zinc oxide NanoArc™ ZN-2605
Show More Show Less
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 215-222-5
RTECSNumber ZH4810000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only