missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Oxide, Honeywell Fluka™
Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5% (calc. for dried substance)
$588.18 - $588.18
Chemical Identifiers
| CAS | 1314-13-2 |
|---|---|
| Molecular Formula | OZn |
| Molecular Weight (g/mol) | 81.38 |
| MDL Number | MFCD00011300 |
| InChI Key | XLOMVQKBTHCTTD-UHFFFAOYSA-N |
| Synonym | zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 |
| PubChem CID | 14806 |
| ChEBI | CHEBI:36560 |
| IUPAC Name | oxozinc |
| SMILES | O=[Zn] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1314-13-2 | |
| 81.38 | |
| XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
| 14806 | |
| oxozinc |
| OZn | |
| MFCD00011300 | |
| zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
| CHEBI:36560 | |
| O=[Zn] |
Specifications
| 1314-13-2 | |
| OZn | |
| MFCD00011300 | |
| zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 | |
| O=[Zn] | |
| 81.38 | |
| CHEBI:36560 | |
| 99 to 100.5% (calc. to the dried substance) | |
| Zinc Oxide, Meets analytical specification of Ph. Eur.BP, USP |
| 1 kg | |
| ZnO | |
| 3077 | |
| XLOMVQKBTHCTTD-UHFFFAOYSA-N | |
| oxozinc | |
| 14806 | |
| 81.39g/mol | |
| puriss. |
Safety and Handling
H410
EINECSNumber : 215-222-5