missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, heavy, 98%

CAS: 1314-13-2 | OZn | 81.38 g/mol

Supplier:  Thermo Scientific Chemicals A161880B

 View more versions of this product

Catalog No. AAA161880B


Only null left
Explore available promotions

Description

Description

Mainly used in coating auxiliary agents, electronics chemicals, leather auxiliary agents, paper chemicals, petroleum additives, plastic auxiliary agents, rubber auxiliary agents,surfactants, textile auxiliary agents. Nanometer zinc oxide can also be used for high-grade ceramic material, can reduce the sintering temperature, achieving the purpose of saving energy. Used as the sulfuration active agents and reinforcing agents for various natural rubber and compound rubber products as well as the colorants and filling agents of white glue. It can also be used as additives to improve textile, textile ultraviolet resistant function and self-cleaning ability, can also be used in feed industry additive

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly used in coating auxiliary agents, electronics chemicals, leather auxiliary agents, paper chemicals, petroleum additives, plastic auxiliary agents, rubber auxiliary agents,surfactants, textile auxiliary agents. Nanometer zinc oxide can also be used for high-grade ceramic material, can reduce the sintering temperature, achieving the purpose of saving energy. Used as the sulfuration active agents and reinforcing agents for various natural rubber and compound rubber products as well as the colorants and filling agents of white glue. It can also be used as additives to improve textile, textile ultraviolet resistant function and self-cleaning ability, can also be used in feed industry additive

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong alkaline earth metal. Alkali metals.
Specifications

Specifications

Zinc oxide
heavy
1000 g
MFCD00011300
14,10147
Insoluble in water.
O=[Zn]
81.38
CHEBI:36560
98%
5.606
1975°C
1314-13-2
OZn
UN3077
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.37
Odorless

Chemical Identifiers

1314-13-2
81.38
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 215-222-5

RTECSNumber : ZH4810000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only