missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc oxide, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1975.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 kg |
| Linear Formula | ZnO |
| Fieser | 01,1294; 05,622 |
| Merck Index | 15,10347 |
| Solubility Information | Solubility in water: 1.6 mg/l (29°C). Other solubilities: soluble in acids and alkalies, soluble in diluted acetic or mineral acids, ammoni, ammonium carbonate, fixed alkali hydroxide solutio, insoluble in alcohol |
| Formula Weight | 81.38 |
| Percent Purity | ≥99.0% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Avoid release to the environment. |
| EINECSNumber | 215-222-5 |
RUO – Research Use Only