missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide, ACS,Reag. Ph Eur, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1088499025
Change view
Click to view available options
Quantity:
500 g
1 kg
25 kg
Packaging:
Fibre carton
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1088499025 25 kg Fibre carton
M1088490500 500 g Plastic bottle
M1088491000 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1088499025 Supplier Technidata Supplier No. 1.08849.9025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For analysis

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Physical Form Solid
Quantity 25 kg
UN Number UN 3077
Formula Weight 81.37. (molar mass)
Grade ACS/Reagent Ph. Eur.
Odor Odorless
Packaging Fibre carton
Appearance White solid
Chemical Name or Material Zinc oxide
Hazard Category H410: Very toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 215-222-5