missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, 99.999%, (trace metal basis)

Catalog No. AC315790250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC315790250 25 g Glass bottle
AC315791000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC315790250 Supplier Thermo Scientific Chemicals Supplier No. 315790250

CAS: 1314-13-2 | OZn | 81.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975.0°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.999%
Linear Formula ZnO
Fieser 01,1294; 05,622
Merck Index 15, 10347
Solubility Information Solubility in water: 1.6 mg/l (29°C). Other solubilities: soluble in acids and alkalies, soluble in diluted acetic or mineral acids, ammoni, ammonium carbonate, fixed alkali hydroxide solutio, insoluble in alcohol
Formula Weight 81.38
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 5.6
Density 5.6000g/mL
Chemical Name or Material Zinc oxide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
EINECSNumber 215-222-5

RUO – Research Use Only