missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Nitrate, Crystal, 98.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystals or Needles |
| Quantity | 2.5 kg |
| Molecular Weight (g/mol) | 297.47 |
| Percent Purity | 98.5% |
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
| Appearance | To Pass Test |
| Chemical Name or Material | Zinc Nitrate, Crystal |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.