missing translation for 'onlineSavingsMsg'
Learn More

Zinc Nitrate, Crystal, 98.5%, Spectrum™ Chemical

Catalog No. 18614780
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614780 25 g Amber Glass Bottle
18614778 125 g Amber Glass Bottle
18614781 500 g Amber Glass Bottle
18614783 2.5 kg Amber Glass Bottle
18614779 12 kg Poly Pail
18614782 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18614780 Supplier Spectrum Chemical Mfg Cor Supplier No. Z107525GM

Chemical Identifiers

CAS 10196-18-6

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98.5%
Grade Reagent
Packaging Amber Glass Bottle
Appearance To Pass Test

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.