missing translation for 'onlineSavingsMsg'
Learn More

Zinc mossy, 2.5cm (0.98in) & down, 99% (metals basis)

Catalog No. AA14629A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14629A1 1 kg
AA1462930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA14629A1 Supplier Thermo Scientific Chemicals Supplier No. 014629A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-66-6 | Zn | 65.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Quantity 1 kg
Odor Odorless
Particle Size 2.5cm (0.98 in.) & down
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Zinc mossy
EINECSNumber 231-175-3
RTECSNumber ZG8600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only