missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Metal Powder (Dust), Reagents
Specifications
Specifications
| Color | Silver-Gray |
| Boiling Point | 907°C |
| Avg. Mol. Wt. or Mol. Wt. Range | 65.38 |
| Quantity | 3 kg |
| Packaging | Poly Bottle |
| Form | Solid |
| Chemical Name or Material | Zinc Metal Powder (Dust) |